missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P3H2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15723-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the prolyl 3-hydroxylase subfamily of 2-oxo-glutarate-dependent dioxygenases. These enzymes play a critical role in collagen chain assembly, stability and cross-linking by catalyzing post-translational 3-hydroxylation of proline residues. Mutations in this gene are associated with nonsyndromic severe myopia with cataract and vitreoretinal degeneration, and downregulation of this gene may play a role in breast cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Especificaciones
P3H2 | |
Polyclonal | |
Unconjugated | |
P3H2 | |
leprecan like 1, Leprecan like protein 1, LEPREL1, MLAT4, P3H2, Prolyl 3 hydroxylase 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
210530, 55214 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CG71, Q8IVL5 | |
P3H2 | |
P3H2 Fusion Protein Ag8444 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |