missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ORM1 (Human) Recombinant Protein
Human ORM1 full-length ORF ( AAH26238, 18 a.a. - 201 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005004-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: AQIPLCANLVPVPITNATLDRITGKWFYIASAFRNEEYNKSVQEIQATFFYFTPNKTEDTIFLREYQTRQDQCIYNTTYLNVQRENGTISRYVGGQEHFAHLLILRDTKTYMLAFDVNDEKNWGLSVYADKPETTKEQLGEFYEALDCLRIPKSDVVYTDWKKDKCEPLEKQHEKERKQEEGES
Especificaciones
AAH26238 | |
orosomucoid 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ORM1 | |
GST |
5004 | |
Wheat germ expression system | |
25 μg | |
AGP-A, AGP1, ORM | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |