missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPEPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17211-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Probably catalyzes the removal of unsubstituted N-terminal amino acids from various peptides.Especificaciones
NPEPL1 | |
Polyclonal | |
Unconjugated | |
NPEPL1 | |
aminopeptidase like 1, KIAA1974, NPEPL1, Probable aminopeptidase NPEPL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
228961, 311671, 79716 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6NSR8, Q8NDH3 | |
Npepl1 | |
NPEPL1 Fusion Protein Ag10994 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |