missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19953-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody recognize the C-term of GRIN2A.
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Especificaciones
NMDAR2A | |
Polyclonal | |
Unconjugated | |
GRIN2A | |
GluN2A, GRIN2A, hNR2A, NMDAR2A, NR2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14811, 2903 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35436, Q12879 | |
Grin2a | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |