missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKIRAS2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10174-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Atypical Ras-like protein that acts as a potent regulator of NF-kappa-B activity by preventing the degradation of NF-kappa-B inhibitor beta (NFKBIB) by most signals, explaining why NFKBIB is more resistant to degradation. May act by blocking phosphorylation of NFKBIB and nuclear localization of p65/RELA NF-kappa-B subunit. It is unclear whether it acts as a GTPase. Both GTP- and GDP-bound forms block phosphorylation of NFKBIB.Especificaciones
NKIRAS2 | |
Polyclonal | |
Unconjugated | |
NKIRAS2 | |
Kappa B Ras protein 2, kappaB Ras2, KBRAS2, NKIRAS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28511 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NYR9 | |
NKIRAS2 | |
NKIRAS2 Fusion Protein Ag0221 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |