missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NDUFA13 Recombinant Protein
Marca: Abnova™ H00051079-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH09189 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
41.58 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MAASKVKQDMPPPGGYGPIDYKRNLPRRGLSGYSMLAIGIGTLIYGHWSIMKWNRERRRLQIEDFEARIALLPLLQAETDRRTLQMLRENLEEEAIIMKDVPDWKVGESVFHTTRWVPPLIGELYGLRTTEEALHASHGFMWYT | |
B16.6/CDA016/CGI-39/GRIM-19/GRIM19 | |
NDUFA13 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
51079 | |
GRIM19 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NDUFA13 | |
Human | |
Recombinant | |
Solution |