missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NBS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55025-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody is specific to NBN.
The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.Especificaciones
NBS1 | |
Polyclonal | |
Unconjugated | |
NBN | |
AT V1, AT V2, ATV, NBN, NBS, NBS1, nibrin, P95 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27354, 4683, 85482 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60934, Q9JIL9, Q9R207 | |
NBN | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |