Learn More
MYO9A Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26869-1-AP-150UL
Detalles adicionales : Peso : 0.00020kg
Descripción
The MYO9A antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MYO9A. This antibody recognizes human, mouse antigen. The MYO9A antibody has been validated for the following applications: IHC,ELISA analysis.
Especificaciones
MYO9A | |
Polyclonal | |
Unconjugated | |
150 μL | |
4649 | |
Liquid |
Immunohistochemistry, ELISA | |
500 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |