missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYLK2 (skMLCK) Recombinant Human Protein
MYLK2 (skMLCK) is a calcium/calmodulin-dependent serine/threonine kinase.
Marca: Invitrogen™ PV4025
Detalles adicionales : Peso : 0.00900kg
Descripción
MYLK2 (skMLCK) is expressed exclusively in adult skeletal muscle.
Especificaciones
NP_149109 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
MYLK2 (skMLCK) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
92.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MLCK Kinase Family | |
Recombinant | |
Baculovirus |