missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15354-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Myosin, a structural component of muscle, consists of two heavy chains and four light chains. Myosin light chain may regulate muscle contraction by modulating the ATPase activity of myosin heads. It also binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found.Especificaciones
MYL9 | |
Polyclonal | |
Unconjugated | |
MYL9 | |
20 kDa myosin light chain, LC20, MLC 2C, MLC2, MRLC1, MYL9, Myosin RLC, MYRL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10398, 296313, 98932 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24844, Q64122, Q9CQ19 | |
MYL9 | |
MYL9 Fusion Protein Ag7600 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |