missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTRF1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16694-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain termination codons UAA and UAG.Especificaciones
MTRF1L | |
Polyclonal | |
Unconjugated | |
MTRF1L | |
MRF1L, mtRF1a, MTRF1L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108853, 361473, 54516 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4V7E5, Q8BJU9, Q9UGC7 | |
Mtrf1l | |
MTRF1L Fusion Protein Ag10081 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |