missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTERFD3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26676-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Members of the mTERF (mitochondrial transcription termination factor) family, are mitochondrial proteins that are believed to be transcription termination factors. MTERFD3 was initially identified through differential display analysis and is expressed primarily in heart, liver, pancreas and skeletal muscle. MTERFD3 is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription, and may be a serum-inhibitory factor.Especificaciones
MTERFD3 | |
Polyclonal | |
Unconjugated | |
MTERF2 | |
MTERF domain containing 3, MTERFD3, mTERFL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80298 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q49AM1 | |
MTERF2 | |
MTERFD3 Fusion Protein Ag24704 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |