missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH2 Mouse anti-Human, Clone: 2H5F5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60161-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Plays a role in neurogenesis and neuronal migration By similarity. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.Especificaciones
MSH2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43246 | |
MSH2 | |
MSH2 Fusion Protein Ag7835 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2H5F5 | |
Unconjugated | |
MSH2 | |
COCA1, FCC1, hMSH2, HNPCC, HNPCC1, LCFS2, MSH2, MutS protein homolog 2 | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
4436 | |
-20°C | |
Liquid |