Learn More
MP Biomedicals™ Collagen Type I from Calf Skin
Suitable for cell adhesion in tissue culture as well as gel formation, platelet aggregation and as a substrate for collagenase
Marca: MP Biomedicals™ 0215002683
Detalles adicionales : CAS : 9007-34-5 Peso : 0.00030kg
Descripción
- CAS Number: 9007-34-5
- EC Number: 232-697-4
- MDL Number: MFCD00130825
- Format: Powder
- Soluble in 0.075M sodium citrate pH 3.7 and in dilute acetic acid
For attachment of primary cultures of epithelioid cells and many other cell types
Especificaciones
Tumor Marker | |
250mg | |
Chorionic Gonadotropin from Human Pregnancy Urine | |
Lyophilized |
3.7 | |
Calf Skin | |
-20°C |