missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse PD-L2 / B7-DC Protein, His Tag
350.80€ - 2419.31€
Especificaciones
Número de acceso | NP_067371 |
---|---|
Peso molecular | 24.4 kDa |
Fuente | Mouse PD-L2, His Tag (PD2-M52E3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 219 (Accession # NP_067371). |
Inmunógeno | Leu 20 - Arg 219 |
Requisitos de almacenamiento | -20°C |
Codice del prodotto | Marca | Cantidad | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Cantidad | Prezzo | Quantità e disponibilità | |||||
15887517
|
ACROBiosystems
PD2-M52E3-1MG |
1 mg |
2419.31€
1 miligramo |
Fecha estimada de envío: 08-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15877517
|
ACROBiosystems
PD2-M52E3-100UG |
100 μg |
350.80€
100 microgramos |
Fecha estimada de envío: 08-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descrizione
Programmed cell death 1 ligand 2 (PD-L2 or PDCD1 ligand 2) is also known as Butyrophilin B7-DC, CD antigen CD273, which belongs to the immunoglobulin superfamily or BTN/MOG family. The expression of PD-L2 is up-regulated by IFNG/IFN-gamma stimulation in monocytes and induced on dendritic cells grown from peripheral blood mononuclear cells with CSF2 and IL-4. PD-L2 Involved in the costimulatory signal, essential for T-cell proliferation and IFNG production in a PDCD1-independent manner. PD-L2 interaction with PDCD1 inhibits T-cell proliferation by blocking cell cycle progression and cytokine production.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 37-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
NP_067371 | |
Mouse PD-L2, His Tag (PD2-M52E3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Arg 219 (Accession # NP_067371). | |
-20°C | |
PD-L2 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
24.4 kDa | |
Leu 20 - Arg 219 | |
PDL2,PD-L2,Butyrophilin B7-DC,CD273,PDCD1 ligand 2,PDCD1L2,PDCD1LG2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |