Learn More
ACROBiosystems Mouse IFN-alpha 1 Protein, His Tag
Marca: ACROBiosystems IFA-M52H3-100ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Interferon alpha-1 is also known as IFN-alpha-1/13, Interferon alpha-D, LeIF D and IFNA1, belongs to the alpha / beta interferon family. Interferons alpha-1 and alpha-13 have identical protein sequences. Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase. IFN-alpha can either suppress or promote the development of autoimmune diabetes. It is likely that IFN-alpha plays a complex role in the etiology of type 1 diabetes.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.0 kDa. The protein migrates as 24-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_034632.2 | |
100 μg | |
Cys 24 - Lys 189 | |
IFN-alpha 1,IFNA1,IFNalpha 1,IFN-alpha-1,LeIF D | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
21.0 kDa | |
Mouse IFN-alpha 1, His Tag (IFA-M52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Lys 189 (Accession # P01572-1). | |
-20°C | |
IFN-alpha 1 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
ActiveMax™ Cytokines |