missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MORG1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27244-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MORG1 is a member of the WD-40 protein family. The protein is proposed to function as a molecular scaffold for various multimeric protein complexes. The protein associates with several components of the extracellular signal-regulated kinase (ERK) pathway, and promotes ERK activity in response to serum or other signals. The protein also interacts with egl nine homolog 3 (EGLN3, also known as PHD3) and regulates expression of hypoxia-inducible factor 1, and has been purified as part of the spliceosome.Especificaciones
MORG1 | |
Polyclonal | |
Unconjugated | |
WDR83 | |
MAPK organizer 1, MORG1, WDR83 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67836, 84292 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BRX9, Q9DAJ4 | |
Wdr83 | |
MORG1 Fusion Protein Ag15141 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |