missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIOX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17623-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MIOX (Myo-Inositol Oxygenase) is a Protein Coding gene. Among its related pathways are superpathway of inositol phosphate compounds and Porphyrin and chlorophyll metabolism. Gene Ontology (GO) annotations related to this gene include iron ion binding and oxidoreductase activity, acting on NAD(P)H.Especificaciones
MIOX | |
Polyclonal | |
Unconjugated | |
MIOX | |
Aldehyde reductase like 6, ALDRL6, Inositol oxygenase, Kidney specific protein 32, KSP32, MI oxygenase, MIOX, myo inositol oxygenase, Renal specific oxidoreductase, RSOR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
252899, 55586, 56727 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9QXN4, Q9QXN5, Q9UGB7 | |
MIOX | |
MIOX Fusion Protein Ag11829 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |