Learn More
Kimble Chase™ Micro Separatory Funnels
Kimble Chase™ Micro Separatory Funnels are used in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
Marca: Kimble Chase™ 298280-0010
Detalles adicionales : Peso : 1.00000kg
Descripción
- Standard taper joints
- Squibb-type funnel with a PTFE stopcock plug with a 2mm bore
- Plug is 821001-0002
- Stopper not included
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and STM E438, Type I, Class A requirements
Especificaciones
10 mL | |
141 mm | |
Micro Separatory Funnel, Squibb-type |
Borosilicate Glass | |
ST 14/20 |