Learn More
Cadence Science™ Micro-Mate™ Interchangeable Syringes
Syringe barrels and plungers fit perfectly, eliminating the task of matching up syringe parts
Marca: Cadence Science™ 5025
Detalles adicionales : Peso : 0.01000kg
Descripción
Syringe barrels and plungers fit perfectly, eliminating the task of matching up syringe parts
Interchangeable syringes are manufactured to ensure the perfect mating of barrels and plungers. Any Micro-Mate interchangeable syringe of a particular size will fit any Micro-Mate interchangeable barrel of that same size.
- Metal Male Luer Lock, Metal Male Luer or Glass Male Luer tips
- Constructed of borosilicate glass for heat and corrosion resistance
- Permanently fused graduations Reinforced flanges with two flat sides prevent syringes from rolling
Especificaciones
Interchangeable Syringe | |
10 mL | |
Male Luer Slip |
0.2 mL | |
Micromate Interchangeable Barrel of Same Size |