Learn More
MGP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10734-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MGP is associates with the organic matrix of bone and cartilage. MGP is thought to act as an inhibitor of bone formation.Especificaciones
MGP | |
Polyclonal | |
Unconjugated | |
MGP | |
GIG36, matrix Gla protein, MGLAP, MGP, NTI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4256 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08493 | |
MGP | |
MGP Fusion Protein Ag1091 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |