Learn More
MGLL Mouse, anti-Human, Unconjugated, Clone: 2A3E11, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 67846-1-IG-150UL
Detalles adicionales : Peso : 0.00020kg
 Ver más versiones de este producto
Código de producto. 17201516
  • 349.00€ / 150 microlitros
Fecha estimada de envío: 15-12-2022
para ver el stock
Descripción
Descripción
MGLL (monoglyceride lipase) is also named as MAGL, HU-K5 and belongs to the monoacylglycerol lipase family. It converts monoacylglycerides to free fatty acids and glycerol and functions together with hormone-sensitive lipase (HSL) in the mobilization of fatty acids from the triglyceride stores of adipocytes. MGL protein is ubiquitously expressed in kidney, spleen, heart, liver, stomach, brain, lung, adrenal gland, adipose tissue and in brain,there are two bands of 33 kDa and 35 kDa can be detected. The single protein species expressed in muscle is larger (around 40 kDa) (PMID:11470505). This protein can exsit as a homodimer (PMID:19957260).
Especificaciones
Especificaciones
MGLL
Monoclonal
1000 μg/mL
Mouse
Primary
Pig, Human
ELISA, Western Blot
2A3E11
Unconjugated
150 μL
11343