Learn More
Epredia™ Lab Vision™ Melanoma Marker, Pan Ab-1, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ Melanoma Marker, Pan Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-939-R7
Detalles adicionales : Peso : 0.01200kg
Description
Melanoma associated antigens recognized by autologous cytotoxic T cells include MART-1, MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1.Host Species: Mouse
Clone: HMB45 (gp100), M2-7C10 (MART-1) and M2-9E3 (MART-1)
Isotype: IgG1/κ + IgG2b/κ + IgG2b/κ
Species Reactivity: Human and Horse. Others not known.
Immunogen: Extract of pigmented melanoma metastases from lymph nodes (HMB45); Recombinant hMART-1 protein (M2-7C10 and M2-9E3)
Molecular Weight: 100kDa (gp100) and 18kDa (MART-1)
Positive Control: Melanoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Melanoma Marker, Pan Ab-1 | |
Monoclonal | |
Unconjugated | |
Extract of pigmented melanoma metastases from lymph nodes (HMB45); Recombinant hMART-1 protein (M2-7C10 and M2-9E3) | |
RUO | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
HMB45 (gp100, M2-7C10) MART-1 and M2-9E3 (MART-1) | |
Mouse | |
7 mL | |
Cancer and Tumor Biology | |
Equine | |
IgG2a κ |