missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Melanoma Marker, Pan Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ Melanoma Marker, Pan Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-939-S

Detalles adicionales : Peso : 0.00250kg

 Ver más versiones de este producto

Código de producto. 12672607

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Melanoma associated antigens recognized by autologous cytotoxic T cells include MART-1, MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1.

Host Species: Mouse

Clone: HMB45 (gp100), M2-7C10 (MART-1) and M2-9E3 (MART-1)

Isotype: IgG1/κ + IgG2b/κ + IgG2b/κ

Species Reactivity: Human and Horse. Others not known.

Immunogen: Extract of pigmented melanoma metastases from lymph nodes (HMB45); Recombinant hMART-1 protein (M2-7C10 and M2-9E3)

Molecular Weight: 100kDa (gp100) and 18kDa (MART-1)

Positive Control: Melanoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Melanoma Marker, Pan Ab-1
Monoclonal
Unconjugated
Extract of pigmented melanoma metastases from lymph nodes (HMB45); Recombinant hMART-1 protein (M2-7C10 and M2-9E3)
1 mL
Cancer and Tumor Biology
Equine, Human
IgG2a κ
Immunohistochemistry (Paraffin)
HMB45 (gp100, M2-7C10) MART-1 and M2-9E3 (MART-1)
Mouse
Melanoma
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales