missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCCC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14861-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MCCC1 is the large subunit of3-methylcrotonyl-CoA carboxylase. This enzyme functions as a heterodimer and catalyzes the carboxylation of 3-methylcrotonyl-CoAto form 3-methylglutaconyl-CoA.Especificaciones
MCCC1 | |
Polyclonal | |
Unconjugated | |
MCCC1 | |
MCC B, MCCA, MCCase subunit alpha, MCCC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294972, 56922, 72039 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5I0C3, Q96RQ3, Q99MR8 | |
Mccc1 | |
MCCC1 Fusion Protein Ag6663 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |