Learn More
MBTD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26098-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MBDT1 is a putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility. MBDT1 specifically binds to monomethylated and dimethylated ′Lys-20′ on histone H4.Especificaciones
MBTD1 | |
Polyclonal | |
Unconjugated | |
MBTD1 | |
mbt domain containing 1, MBTD1, SA49P01 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103537, 54799 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q05BQ5, Q6P5G3 | |
MBTD1 | |
MBTD1 Fusion Protein Ag22059 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |