missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBIP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10685-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MBIP inhibits the MAP3K12 activity to induce the activation of the JNK/SAPK pathway. Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.Especificaciones
MBIP | |
Polyclonal | |
Unconjugated | |
MBIP | |
MBIP, MUK binding inhibitory protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51562 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NS73 | |
MBIP | |
MBIP Fusion Protein Ag1061 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |