missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBIP Mouse anti-Human, Clone: 2E6H11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66102-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
MBIP inhibits the MAP3K12 activity to induce the activation of the JNK/SAPK pathway. Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.Especificaciones
MBIP | |
Monoclonal | |
1.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NS73 | |
MBIP | |
MBIP Fusion Protein Ag18090 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
2E6H11 | |
Unconjugated | |
MBIP | |
MBIP, MUK binding inhibitory protein | |
Mouse | |
Protein A | |
RUO | |
51562 | |
-20°C | |
Liquid |