Learn More
Abnova™ MATP Recombinant Protein
Human MATP full-length ORF ( AAH03597, 1 a.a. - 243 a.a.) recombinant protein with GST-tag at N-terminal
Marca: Abnova™ H00051151-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a transporter protein that mediates melanin synthesis. The protein is expressed in a high percentage of melanoma cell lines. Mutations in this gene are a cause of oculocutaneous albinism type 4, and polymorphisms in this gene are associated with variations in skin and hair color. Multiple transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)
- Molecular weight: 52.47kDa
- Preparation method: in vitro wheat germ expression system
- Purification:Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8 in the elution buffer
- Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue
Best use within three months from the date of receipt of this protein
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH03597 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
52.47 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MGSNSGQAGRHIYKSLADDGPFDSVEPPKRPTSRLIMHSMAMFGREFCYAVEAAYVTPVLLSVGLPSSLYSIVWFLSPILGFLLQPVVGSASDHCRSRWGRRRPYILTLGVMMLVGMALYLNGATVVAALIANPRRKLVWAISVTMIGVVLFDFAADFIDGPIKAYLFDVCSHQDKEKGLHYHALFTDSQGNDIKVTAESTGEHASSLPLPLHQPPHWMDGLPVQHAVLHRFHGPDCVPRGSL | |
1A1/AIM1/MATP/SHEP5 | |
SLC45A2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
51151 | |
MATP (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SLC45A2 | |
Human | |
Recombinant | |
Solution |