missing translation for 'onlineSavingsMsg'
Learn More

MAPK13 Mouse anti-Human, Mouse, Clone: 1B12A6, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60252-1-IG-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16827295

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 04-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and cellular stress. MAP kinase kinases 3, and 6 can phosphorylate and activate this kinase. Transcription factor ATF2, and microtubule dynamics regulator stathmin have been shown to be the substrates of this kinase.
Especificaciones

Especificaciones

MAPK13
Monoclonal
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15264, Q9Z1B7
MAPK13
MAPK13 Fusion Protein Ag21240
20 μL
Primary
Human, Mouse
Antibody
IgG1
Western Blot
1B12A6
Unconjugated
MAPK13
MAP kinase 13, MAP kinase p38 delta, MAPK 13, MAPK13, p38delta, PRKM13, SAPK4
Mouse
Protein G
RUO
26415, 5603
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales