missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP4K5 (KHS1) Recombinant Human Protein
MAP4K5 (KHS1) is a serine/threonine kinase that belongs to the GCK family of kinases.
Marca: Invitrogen™ PV4014
Detalles adicionales : Peso : 0.00900kg
Descripción
MAP4K5 (KHS1) has been implicated as an upstream regulator of MAP kinase signaling pathways.
Especificaciones
NP_942089 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
MAP4K5 (KHS1) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
122.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |