missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LGALS8 (Human) Recombinant Protein
Human LGALS8 full-length ORF ( AAH15818, 1 a.a. - 358 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003964-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MLSLNNLQNIIYNPVIPYVGTIPDQLDPGTLIVICGHVPSDADRFQVDLQNGSSVKPRADVAFHFNPRFKRAGCIVCNTLINEKWGREEITYDTPFKREKSFEIVIMVLKDKFQVAVNGKHTLLYGHRIGPEKIDTLGIYGKVNIHSIGFSFSSDLQSTQASSLELTEISRENVPKSGTPQLPSNRGGDISKIAPRTVYTKSKDSTVNHTLTCTKIPPMNYVSKSLPFAARLNTPMGPGRTVVVKGEVNANAKSFNVDLLAGKSKDIALHLNPRLNIKAFVRNSFLQESWGEEERNITSFPFSPGMYFEMIIYCDVREFKVAVNGVHSLEYKHRFKELSSIDTLEINGDIHLLEVRSW
Especificaciones
AAH15818 | |
lectin, galactoside-binding, soluble, 8 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LGALS8 | |
GST |
3964 | |
Wheat germ expression system | |
25 μg | |
Gal-8, PCTA-1, PCTA1, Po66-CBP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |