Learn More
Invitrogen™ LanthaScreen™ Amine Reactive Tb Chelate
LanthaScreen™ Amine Reactive Tb Chelate
494.00€ - 17870.00€
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
11563467
|
Invitrogen™
PV3583 |
10 μg |
494.00€
10 microgramos |
Fecha estimada de envío: 27-05-2024
Inicie sesión para ver el stock |
|||||
11553467
|
Invitrogen™
PV3582 |
100 μg |
2405.00€
100 microgramos |
Fecha estimada de envío: 27-05-2024
Inicie sesión para ver el stock |
|||||
10595343
|
Invitrogen™
PV3581 |
1 mg |
17870.00€
1 miligramo |
Fecha estimada de envío: 27-05-2024
Inicie sesión para ver el stock |
|||||
Descripción
As part of the LanthaScreen™ TR-FRET toolbox of assay reagents, LanthaScreen™ Amine Reactive Tb Chelate labeling reagent is available for assay development. The amine-reactive, isothiocyanate group conjugates to virtually any peptide or protein that contains one or more accessible amine moieties, such as the amine group found on a lysine residue or an unmodified N-terminal amine group of a peptide or protein. Amine-modified DNA oligonucleotides can be labeled as well. Energy transfer from the terbium donor to a suitable acceptor such as fluorescein is readily detected by monitoring an increase in acceptor fluorescence intensity. See the user guide for application-based protocols for labeling peptides containing free amino groups, amine-modified oligonucleotides, and IgG antibodies.For Research Use Only. Not for use in diagnostic procedures.