Learn More
Epredia™ Lab Vision™ Tubulin-Β Ab-6 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Epredia™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marca: Epredia™ MS-1226-P
Detalles adicionales : Peso : 0.00100kg
Descripción
Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.Host Species: Mouse
Clone: TBN06; same as Tub 2.5
Isotype: IgG3
Species Reactivity: Human, Cow, Rabbit, Mouse, Chicken, Rat, Frog, Moth and Wheat. Others not known.
Epitope: Not determined
Immunogen: Purified tubulin
Molecular Weight: 55kDa
Positive Control: Ls174T, MAD109 cells; Skin or lung
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Especificaciones
Tubulin-beta Ab-6 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Cytoskeletal Protein | |
Bovine, Chicken, Frog, Human, Moth, Mouse, Rabbit, Rat, Wheat | |
IgG3 |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
TBN06; same as Tub 2.5 | |
Unconjugated | |
Purified tubulin | |
RUO | |
Primary | |
Concentrated |