missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Tubulin-Β Ab-6 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Epredia™ Tubulin-Β Ab-6, Mouse Monoclonal Antibody provides accurate, reproducible results.

Marca:  Epredia™ MS-1226-R7

Detalles adicionales : Peso : 0.01200kg

 Ver más versiones de este producto

Código de producto. 12669326

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.

Host Species: Mouse

Clone: TBN06; same as Tub 2.5

Isotype: IgG3

Species Reactivity: Human, Cow, Rabbit, Mouse, Chicken, Rat, Frog, Moth and Wheat. Others not known.

Epitope: Not determined

Immunogen: Purified tubulin

Molecular Weight: 55kDa

Positive Control: Ls174T, MAD109 cells; Skin or lung

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Tubulin-beta Ab-6
Monoclonal
Unconjugated
Purified tubulin
RUO
Primary
Prediluted
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
TBN06; same as Tub 2.5
Mouse
7 mL
Cytoskeletal Protein
Bovine, Chicken, Frog, Human, Moth, Mouse, Rabbit, Rat, Wheat
IgG3
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales