Learn More
Epredia™ Lab Vision™ Thomsen-Friedenreich Antigen Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Provide accurate, reproducible results with the Epredia™ Thomsen-Friedenreich Antigen Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-455-P0
Detalles adicionales : Peso : 1.00000kg
Descripción
The Thomsen-Fridenreich antigen acts as an oncofetal antigen, with low expression in normal adult tissues but increasing to fetal levels of expression in hyperplasia or malignancy. It is considered a pan-carcinoma marker.
Host Species: Mouse
Clone: A78-G/A7
Isotype: IgM/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: Galβ1-3GalNAc
Immunogen: Neuraminidase-treated human red blood cells
Positive Control: KG1 cells, Human colorectal CA tissues
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Especificaciones
Thomsen-Friedenreich Antigen Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Cancer and Tumor Biology | |
Human, Rat | |
IgM κ |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
A78-G/A7 | |
Unconjugated | |
Neuraminidase-treated human red blood cells | |
RUO | |
Primary | |
Concentrated |