Learn More
Epredia™ Lab Vision™ Retinoblastoma (Rb) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Provide accurate, reproducible results with the Epredia™ Retinoblastoma (Rb) Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-107-P
Detalles adicionales : Peso : 0.00250kg
Descripción
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia.Host Species: Mouse
Clone: 1F8; same as Rb1
Isotype: IgG1
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: aa 703-722
Immunogen: Recombinant human Rb protein
Molecular Weight: 105kDa
Positive Control: Ls174T, HT29, HUVEC cells; Normal colon, Breast CA and colon CA
Cellular Localization: Nuclear Ab-1 specifically stains the nuclei of BT-20 cells and primary human foreskin fibroblast (HFF) cells. It reacts with hyperphosphorylated and un (under) phosphorylated form of Rb protein. It shows no cross reaction with p107 and p130.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Especificaciones
Retinoblastoma (Rb) Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Cancer and Tumor Biology | |
Human | |
IgG1 |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1F8; same as Rb1 | |
Unconjugated | |
Recombinant human Rb protein | |
RUO | |
Primary | |
Concentrated |