Learn More
Epredia™ Lab Vision™ Progesterone Receptor Rabbit Monoclonal Antibody
Progesterone Receptor, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
Marca: Epredia™ RM-9102-S0
Detalles adicionales : Peso : 4.98950kg
Description
The progesterone receptor (PgR) is an estrogen-regulated protein. It has been proposed that expression of PgR determination indicates a responsive estrogen receptor (ER) pathway, and therefore, may predict likely response to endocrine therapy in human breast cancer. A number of studies have shown that PgR determination provides supplementary information to ER, both in predicting response to endocrine therapy and estimating survival. PgR has proved superior to ER as a prognostic indicator in some studies.Host Species: Rabbit
Clone: SP2
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: 412-526aa
Immunogen: Recombinant protein encoding human progesterone receptor 412-526aa
Molecular Weight: 116 + 81kDa
Positive Control: Breast CA, T47D cells
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Progesterone Receptor (SP2) | |
Monoclonal | |
Unconjugated | |
Recombinant protein encoding human progesterone receptor 412-526 aa | |
IVD | |
Primary | |
Concentrated |
Western Blot | |
SP2 | |
Rabbit | |
100 μL | |
Cancer and Tumor Biology | |
Human | |
IgG |