missing translation for 'onlineSavingsMsg'
Learn More

Pds1 Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Pds1 Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia MS-1511-R7

Detalles adicionales : Peso : 0.00700kg

 Ver más versiones de este producto

Código de producto. 12629976

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Pds1 is an anaphase inhibitor and plays important role in DNA damage and spindle check point pathways. Pds1 inhibits sister chromatid separation by binding and inhibiting Esp1, cystein protease that causes cleavage of the cohesin Scc1 that binds the sister chromatids together. Pds1 is responsible for targeting of Esp1 to nucleus and its binding to the spindle. Degradation of Pds1 occurs shortly before anaphase, which liberates Esp1 and is a prerequisite for anaphase entry. Pds1 is targeted for degradation by ubiquitination mediated by cyclosome/anaphase-promoting complex (APC) functioning as a ubiquitin ligase. In response to DNA damage Chk1 phosphorylates Pds1 to stabilize it against the APC mediated destruction, hence preventing the entry of such a cell into anaphase.

Host Species: Mouse

Clone: DCS-280

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human recombinant full-length Pds1 protein

Molecular Weight: 42kDa

Positive Control: HeLa cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Pds1 Ab-1
Monoclonal
Unconjugated
Human recombinant full length Pds1 protein
RUO
Primary
Prediluted
Immunohistochemistry (Paraffin), Western Blot
DCS-280
Mouse
7 mL
Cancer and Tumor Biology
Human
IgG2a κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales