missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Pax-5 Rabbit Polyclonal Antibody with BSA and azide

Provide accurate, reproducible results with the Epredia™ Pax-5, Rabbit Polyclonal Antibody.

Marca:  Epredia™ RB-9406-P0

Detalles adicionales : Peso : 0.00170kg

 Ver más versiones de este producto

Código de producto. 12683567

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

PAX-5 is a B cell specific activator protein (BSAP). In the early stages of B cell developmemt, PAX-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20. PAX-5 is expressed primarily inpro-, pre-, and mature B cells, but not in plasma cells. Interestingly, PAX-5 mRNA is transiently detected in the mesencephalon and spinal cord during embryogenesis. Expression then shifts to the fetal liver and correlates with the onset of B lymphopoiesis. This indicates that PAX-5 is important in B cell development, but may also have a role in proper neuronal development.

Host Species: Rabbit

Species Reactivity: Human, Mouse, Rat. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human PAX-5 protein

Molecular Weight: 50kDa

Positive Control: Tonsil, Raji cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Especificaciones

Especificaciones

PAX-5
Polyclonal
Unconjugated
Synthetic peptide derived from C-terminus of human Pax-5 protein
IVD
Primary
Concentrated
Western Blot
200µg/mL
Rabbit
100 μL
B-Cell
Human, Mouse
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales