missing translation for 'onlineSavingsMsg'
Learn More

MMP-2 (72kDa Collagenase IV) Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in experiments with the Epredia™ MMP-2 (72kDa Collagenase IV) Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia MS-567-PABX

Detalles adicionales : Peso : 0.00180kg

 Ver más versiones de este producto

Código de producto. 12641637

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids into an active form.of 62kDa.

Host Species: Mouse

Clone: CA-4001 or CA719E3C

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: N-terminal (APSPIIKFPGD-VAPKTDK)

Immunogen: N-terminal peptide APSPIIKFPGD-VAPKTDK of procollagenase IV

Molecular Weight: 72kDa (proform)

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Not suitable)
  • Inhibition of MMP-2 Activity

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

MMP-2 (72kDa Collagenase IV) Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Proteases And Inhibitors
Human, Mouse, Rat
IgG1
Western Blot
CA-4001 or CA719E3C
Unconjugated
N-terminal peptide APSPIIKFPGD-VAPKTDK of procollagenase IV
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales