missing translation for 'onlineSavingsMsg'
Learn More

HER-2/c-erbB-2/neu Ab-1 Rabbit Polyclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation, Epredia™ HER-2/c-erbB-2/neu Ab-1, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Marca:  Epredia RB-103-P1

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12612797

  • 466.00€ / 500 microlitros
Fecha estimada de envío: 17-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

c-erbB-2, second member (c-erbB-2/HER-2/neu) of the c-erbB family is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Rat and Mouse. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human HER-2/c-erbB-2 protein. This sequence is identical in rat neu protein.

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells, or T47D cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

HER-2/c-erbB-2/neu Ab-1
Polyclonal
Unconjugated
Synthetic peptide derived from C-terminus of human c-erbB-2/HER-2 protein. This Sequence is identical in rat neu protein.
RUO
Primary
Concentrated
Immunoprecipitation, Western Blot
1mg/mL
Rabbit
500 μL
Oncogenesis
Human, Mouse, Non-human Primate
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales