missing translation for 'onlineSavingsMsg'
Learn More

GLUT-1 (SPM498) Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ GLUT-1, Mouse Monoclonal Antibody.

Marca:  Epredia MS-10637-R7

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 12688786

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Glucose is fundamental to the metabolism in mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.

Host Species: Mouse

Clone: SPM498

Isotype: IgG2a/κ

Species Reactivity: Human and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from C-terminal of human GLUT-1

Molecular Weight: 55 kDa

Positive Control: Esophageal CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

GLUT-1
Monoclonal
Unconjugated
A synthetic peptide derived from C-terminal of human GLUT-1
7 mL
Cancer and Tumor Biology
Human, Rat
IgG2a κ
Immunohistochemistry (Paraffin)
SPM498
Mouse
Esophageal CA
IVD
Primary
Prediluted
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales