Learn More
GLUT-1 (SPM498) Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ GLUT-1, Mouse Monoclonal Antibody.
Marca: Epredia MS-10637-P0
Detalles adicionales : Peso : 0.00170kg
Descripción
Glucose is fundamental to the metabolism in mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.Host Species: Mouse
Clone: SPM498
Isotype: IgG2a/κ
Species Reactivity: Human and Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from C-terminal of human GLUT-1
Molecular Weight: 55 kDa
Positive Control: Esophageal CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
GLUT-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
Esophageal CA | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
SPM498 | |
Unconjugated | |
A synthetic peptide derived from C-terminal of human GLUT-1 | |
100 μL | |
Cancer and Tumor Biology | |
Human, Rat | |
IgG2a κ |