Learn More
Epithelial Membrane Antigen/EMA Ab-3 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Membrane Antigen/EMA Ab-3, Mouse Monoclonal Antibody.
Marca: Epredia MS-741-R7
Detalles adicionales : Peso : 0.01200kg
Descripción
Epithelial membrane antigen (EMA), is a mucin-like glycoprotein. Antibody to EMA has been shown useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in the bone marrow or liver.Host Species: Mouse
Clone: E29
Isotype: IgG2a/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: RPAP
Immunogen: Delipidated extract of human milk fat globule membranes
Molecular Weight: 265-400 kDa
Positive Control: Breast or Colon CA
Cellular Localization: Cytoplasmic and Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
Epithelial Membrane Antigen/EMA Ab-3 | |
Monoclonal | |
— | |
Mouse | |
7 mL | |
Oncogenesis | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
E29 | |
Unconjugated | |
Delipidated extract of human milk fat globule membranes | |
IVD | |
Primary | |
Prediluted |