missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-13 Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunoprecipitation experiments with Epredia™ Epidermal Growth Factor Receptor/EGFR Ab-13, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-609-B

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12661747

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 528 + 199.12

Isotype: IgG2a + IgG2a

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Extracellular domain

Immunogen: Partially purified EGFR from A431 cells (clone 528) and extracellular domain of recombinant human EGFR protein (clone 199.12)

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Epidermal Growth Factor Receptor/EGFR Ab-13
Monoclonal
200μg/mL
Mouse
A431 cells
RUO
Primary
Concentrated
Immunoprecipitation
528 + 199.12
Biotin
Partially purified EGFR from A431 cells (clone 528) and extracellular domain of recombinant human EGFR protein (clone 199.12)
1 mL
EGFR Family
Human
IgG2a
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales