missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 19 Ab-5 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and western blotting procedures with Epredia™ Cytokeratin 19 Ab-5, Mouse Monoclonal Antibody.

Marca:  Epredia MS-1902-P1ABX

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12630437

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues except in liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: RCK108

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Cytoskeleton preparation of T24 cells

Molecular Weight: 40 kDa

Positive Control: LS174T and HT29 cells, Colon CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Cytokeratin 19 Ab-5
Monoclonal
1mg/mL
Mouse
100 μL
Cytoskeletal Protein
Human
IgG1
Immunohistochemistry (Paraffin), Western Blot
RCK108
Unconjugated
Cytoskeleton preparation of T24 cells
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales