Learn More
Epredia™ Lab Vision™ CD42b (GPIb) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-1174-P0
Detalles adicionales : Peso : 0.00170kg
Descripción
The CD42b glycoprotein, also known as GPIb, is a cofactor of ristocetin-induced aggregation and is involved in the binding of platelets to blood vessel walls. The CD42b antigen is expressed on platelets and on megakaryocytes in bone marrow. The absence of CD42b antigen on platelets may indicate Bernard-Soulier disease.Host Species: Mouse
Clone: 42C01
Isotype: IgG1
Species Reactivity: Human. Others not tested.
Immunogen: Recombinant CD42b.
Molecular Weight: 135-145 kDa (α-chain) and 22-25 kDa (β-chain)
Positive Control: Tonsil
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Especificaciones
CD42b Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Hematopoietic | |
Human | |
IgG1 |
Immunohistochemistry (Paraffin) | |
42C01 | |
Unconjugated | |
Recombinant CD42b | |
RUO | |
Primary | |
Concentrated |