Learn More
Calprtectin-Monocyte/Macrophage/L1 protein Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ Calprtectin-Monocyte/Macrophage/L1 protein Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marca: Epredia MS-148-P1
Detalles adicionales : Peso : 1.00000kg
Descripción
Host Species: Mouse
Clone: MAC 387
Isotype: IgG1
Species Reactivity: Human, Baboon, Monkey, Pig, Cat, Dog, Guinea pig, Rat, and Mouse. Others not known.
Epitope: Not determined
Immunogen: Affinity purified monocyte membrane preparation
Positive Control: Tonsil, lymph node, or spleen
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Especificaciones
Macrophage/L1 Protein/Calprotectin Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Hematopoietic | |
Canine, Feline, Guinea Pig, Human, Mouse, Non-human Primate, Porcine, Rat | |
IgG1 |
Immunohistochemistry (Paraffin) | |
MAC 387 | |
Unconjugated | |
Affinity purified monocyte membrane preparation | |
IVD | |
Primary | |
Concentrated |