missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Calponin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Calponin Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-1168-PABX

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12699176

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Calponin, a calmodulin, binds tropomyosin and F-actin and is thought to be involved in the regulation of smooth muscle contraction. The expression of calponin is restricted to smooth muscle cells and is a marker of the differentiated contractile phenotype of developing smooth muscle. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. Expression of the 29 kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions.

Host Species: Mouse

Clone: CALP

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not tested.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 34 kDa

Positive Control: Myoepithelial cells in breast ducts

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

Calponin Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Muscle Biology
Human, Rat
IgG2a κ
Immunohistochemistry (Paraffin)
CALP
Unconjugated
Crude human uterus extract
IVD
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales